Learn More
Ingegrin αvβ5 Mouse, Alexa Fluor 647, Clone: ALULA, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565836
Description
The ALULA monoclonal antibody specifically binds to Integrin alpha v beta 5 ( α v β5 ), which is also known as CD51/β5 . Integrin αvβ5 is widely expressed by fibroblasts, endothelial cells, and vascular smooth muscle cells. It is involved in activation-dependent cell migration and cell adhesion to t he extracellular matrix. Integrin αvβ5 play s a major role in growth factor-induced angiogenesis in response to VEGF and TGF. The αv Integrins, αvβ5 and αvβ3 β5 integrin and functionally block s the interactio n between α v β5 and vitronectin. Although raised against mouse α v β5 , the ALULA antibody crossreacts with human α v β5 .

Specifications
Ingegrin αvβ5 | |
Monoclonal | |
0.2mg/mL | |
Integrin alpha v beta 5; αVβ5; CD51/β5 | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG2b κ |
Flow Cytometry | |
ALULA | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse fibroblasts expressing αvβ5 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.