Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INMT, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106479 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-479 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-479 Supplier Abnova Corporation Supplier No. H00011185A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant INMT.

N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine. [provided by RefSeq

Sequence: DAYRAALCNLASLLKPGGHLVTTVTLRLPSYMVGKREFSCVALEKGEVEQAVLDAGFDIEQLLHSPQSYSVTNAANNGVCCIVARKKPGP

Specifications

Antigen INMT
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant INMT.
Formulation 50% glycerol
Gene INMT
Gene Accession No. NM_006774
Gene Alias MGC125940/MGC125941
Gene Symbols INMT
Host Species Mouse
Immunogen INMT (NP_006765, 174 a.a. ∽ 263 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11185
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.