Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INS, Mouse, Biotin, Clone: D3E7, Abnova™

Catalog No. 89112253 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-253 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-253 Supplier Abnova Corporation Supplier No. MAB5443

Mouse monoclonal antibody raised against INS.

After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds. Binding of this mature form of insulin to the insulin receptor (INSR) stimulates glucose uptake. A variety of mutant alleles with changes in the coding region have been identified. There is a read-through gene, INS-IGF2, which overlaps with this gene at the 5' region and with the IGF2 gene at the 3' region. [provided by RefSeq]

Specifications

Antigen INS
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone D3E7
Conjugate Biotin
Description insulin
Formulation In 20mM Tris-HCl, 150mM NaCl, pH 7.5 (0.09% sodium azide)
Gene INS
Gene Alias ILPR, IRDN
Gene Symbols INS
Host Species Mouse
Immunogen Purified human INS.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity Cross-reacts with human, bovine and porcine insulin and proinsulin.
Target Species Bovine, Human, Pig
Content And Storage Store at 4°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.