Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INS, Mouse, Clone: 2/5E4, Abnova™

Catalog No. 89111534 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-534 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-534 Supplier Abnova Corporation Supplier No. MAB3684
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant INS.

After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds. Binding of this mature form of insulin to the insulin receptor (INSR) stimulates glucose uptake. A variety of mutant alleles with changes in the coding region have been identified. There is a read-through gene, INS-IGF2, which overlaps with this gene at the 5' region and with the IGF2 gene at the 3' region. [provided by RefSeq]

Specifications

Antigen INS
Applications ELISA
Classification Monoclonal
Clone 2/5E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant INS.
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS
Gene INS
Gene Alias ILPR/IRDN
Gene Symbols INS
Host Species Mouse
Immunogen Recombinant protein corresponding to human INS.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity This antibody recognizes natural insulin.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.