Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INS, Mouse, Clone: E2-E3, Abnova™

Catalog No. 89112163 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-163 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-163 Supplier Abnova Corporation Supplier No. MAB5201
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against INS.

Specifications

Antigen INS
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone E2-E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against INS.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene INS
Gene Symbols INS
Host Species Mouse
Immunogen Pig insulin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397415
Test Specificity Insulin is a protein consisting of an alpha-chain of 21 amino acids and a beta-chain of 30 amino acids and produced in the beta-cells of the pancreas. E2-E3 is a specific insulin antibody as tested by ELISA and on human pancreatic tissue. The mouse spleen cells were fused with SP2/0 Ag14 cells and the clones were selected on anti-insulin activity.
Target Species Bovine, Human, Pig
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.