Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin, Mouse, Clone: IN-05, Abnova™

Catalog No. 89135523 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-523 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-523 Supplier Abnova Corporation Supplier No. MAB3863
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native insulin.

Specifications

Antigen Insulin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone IN-05
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native insulin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified porcine insulin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with insulin, one of the major regulatory endocrine hormones of intermediate metabolism, normally secreted by the beta cells (a type of islet cells) of the pancreas; it is also present in tumors of B cell origin such as insulinoma.
Target Species Bovine, Human, Pig
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.