Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RON (CD136) Mouse anti-Human, Unlabeled, Clone: Zt/g4, BD

Catalog No. BDB565691 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565691 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565691 Supplier BD Biosciences Supplier No. 565691
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Zt/g4 monoclonal antibody specifically binds to RON, which is also known as CD136, Macrophage-stimulating protein receptor (MSP-Receptor, MSP-R), Macrophage stimulating 1 receptor (c-met-related tyrosine kinase) (MST1R), or Protein-tyrosine kinase 8 (PTK8). RON is a type I transmembrane glycoprotein that belongs to the receptor tyrosine kinase family. It is expressed on cells as a disulfide-linked heterodimer. RON is expressed on macrophages, keratinocytes, erythroblasts and epithelial cells from the skin, lung, kidney, liver, intestine, colon, and bone marrow. RON reportedly regulates many physiological processes including the survival, migration, differentiation, and effector functions of a variety of cell types including epithelial cells, and macrophages. RON binds Macrophage stimulating protein (MSP). Overexpression of RON may contribute to the malignant transformation of epithelial cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RON (CD136)
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone Zt/g4
Concentration 0.5mg/mL
Conjugate Unconjugated
Description MSP Receptor; MSP-R; MST1R; p185-Ron; PTK8 protein tyrosine kinase 8; PTK8
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human RON Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.