Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin/Proinsulin, Mouse, Clone: D3E7, Abnova™

Catalog No. 89135274 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-274 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-274 Supplier Abnova Corporation Supplier No. MAB2845
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Insulin/Proinsulin.

Specifications

Antigen Insulin/Proinsulin
Applications ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone D3E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Insulin/Proinsulin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Rat and mouse Insulin/Proinsulin.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody cross react with insulin and proinsulin.
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.