Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Integrin alpha6, Clone: 6H7.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABT356
Description
Specifically detects Integrin alpha6 clone: 6H7.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Integrin alpha-6 (UniProt P23229; also known as CD49 antigen-like family member F, CD49f, Integrin alpha6B, VLA-6) is encoded by the ITGA6 (also known as ITGA6B) gene (Gene ID 3655) in human. The biological functions attributed to cell-extracellular matrix (ECM) interactions are mediated primarily by integrin family transmembrane receptors. A total of 18 alpha and 8 beta integrins have been identified in human. Each alpha integrin pairs up with one beta integrin non-covalently to form a functional heterodimeric dimer. At least 24 distinct integrin heterodimers are known with varying ligand specificities. For example, the alpha6-beta1 integrin is reported to mediate the spreading and laminin-binding of Schwann cells, while the alpha6-beta4 integrin is shown to play a role in the suppression of anoikis in human intestinal epithelial cells (IECs). Signal (1-23), Integrin alpha-6 (24-1130). Processed into Integrin alpha-6 heavy chain (24 – 938), Integrin alpha-6 light chain (942-1130), Processed integrin alpha-6 (Alpha6p; 636 – 1130)
Specifications
Integrin alpha6 | |
Monoclonal | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
ITGA6 | |
KLH-conjugated linear peptide corresponding to the cytoplasmic region of human Integrin alpha6 isoforms 2/4/6/7/9. | |
100 μg | |
Cell Structure | |
NP_000201 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
6H7.1 | |
P23229 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction