Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intercalated DNA, Mouse, Clone: 402, Abnova™

Catalog No. 89135641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-641 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-641 Supplier Abnova Corporation Supplier No. MAB4171
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against native Intercalated DNA.

Specifications

Antigen Intercalated DNA
Applications ELISA
Classification Monoclonal
Clone 402
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Intercalated DNA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Ethidium bromide intercalated calf thymus DNA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specifically recognizes intercalated DNA. Does not cross-react with native double-stranded or single-stranded DNA or with ethidium bromide.
Target Species Bovine
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.