Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intra-acrosomal protein, Mouse, Clone: Ds-2, Abnova™

Catalog No. 89135472 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-472 Supplier Abnova Corporation Supplier No. MAB3605
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native intra-acrosomal protein.

Specifications

Antigen Intra-acrosomal protein
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone Ds-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native intra-acrosomal protein.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified canine intra-acrosomal protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with canine intra-acrosomal sperm protein (92 and 42 KDa), atestis-specific acrosomal vesicle protein 1 associated with the acrosomal membranes andmatrix of the mature sperm.
Target Species Canine
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.