Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPO-38, Mouse, Clone: IPO-38, Abnova™

Catalog No. 89135085 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-085 Supplier Abnova Corporation Supplier No. MAB2092
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native IPO-38.

Specifications

Antigen IPO-38
Applications ELISA, Western Blot
Classification Monoclonal
Clone IPO-38
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native IPO-38.
Dilution ELISA (1:1000) Western Blot (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 500mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Mouse
Immunogen Native purified from spleen cells of a human patient with hairy cell leukemia.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.