Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISG15, Mouse, Clone: 3E5, Abnova™

Catalog No. 89109108 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-108 Supplier Abnova Corporation Supplier No. MAB1090
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ISG15.

ISG15 is a ubiquitin-like protein that becomes conjugated to many cellular proteins upon activation by interferon-alpha (IFNA; MIM 147660) and -beta (IFNB; MIM 147640) (Zhao et al., 2005 [PubMed 16009940]).[supplied by OMIM

Specifications

Antigen ISG15
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ISG15.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ISG15
Gene Accession No. NM_005101
Gene Alias G1P2/IFI15/UCRP
Gene Symbols ISG15
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-157 of human ISG15.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9636
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.