Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA1, Mouse, Clone: TS2/7, Abnova™

Catalog No. 89112175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-175 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-175 Supplier Abnova Corporation Supplier No. MAB5224

Mouse monoclonal antibody raised against native ITGA1.

This gene encodes the alpha 1 subunit of integrin receptors. This protein heterodimerizes with the beta 1 subunit to form a cell-surface receptor for collagen and laminin. The heterodimeric receptor is involved in cell-cell adhesion and may play a role in inflammation and fibrosis. The alpha 1 subunit contains an inserted (I) von Willebrand factor type I domain which is thought to be involved in collagen binding. [provided by RefSeq

Specifications

Antigen ITGA1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone TS2/7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ITGA1.
Dilution Flow Cytometry (5 ul of the antibody (neat) to label 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene ITGA1
Gene Alias CD49a/VLA1
Gene Symbols ITGA1
Host Species Mouse
Immunogen Native purified ITGA1 from human thymoma cell line HPB-ALL.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3672
Test Specificity TS2/7 recognizes the human alpha 1 integrin sub-unit, which forms the VLA-1 heterodimer in association with the beta 1 integrin. VLA-1 is a receptor for collagen and laminin, and is expressed by chronically activated T cells, melanoma cells and smooth muscle cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.