Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA11, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106515 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-515 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-515 Supplier Abnova Corporation Supplier No. H00022801A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ITGA11.

This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein contains an I domain, is expressed in muscle tissue, dimerizes with beta 1 integrin in vitro, and appears to bind collagen in this form. Therefore, the protein may be involved in attaching muscle tissue to the extracellular matrix. Alternative transcriptional splice variants have been found for this gene, but their biological validity is not determined. [provided by RefSeq

Sequence: LDARSDLPTAMEYCQRVLRKPAQDCSAYTLSFDTTVFIIESTRQRVAVEATLENRGENAYSTVLNISQSANLQFASLIQKEDSDGSIECVNEERRLQKQVC

Specifications

Antigen ITGA11
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ITGA11.
Formulation 50% glycerol
Gene ITGA11
Gene Accession No. NM_001004439
Gene Alias HsT18964
Gene Symbols ITGA11
Host Species Mouse
Immunogen ITGA11 (NP_001004439, 793 a.a. ∽ 893 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22801
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.