Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2, Mouse, Clone: 244 G1, Abnova™

Catalog No. 89111430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-430 Supplier Abnova Corporation Supplier No. MAB3464
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ITGA2.

This gene product belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane glycoproteins composed of a distinct alpha chain and a common beta chain. They are found on a wide variety of cell types including, T cells, fibroblasts and platelets. Integrins are involved in cell adhesion and also participate in cell-surface mediated signalling. [provided by RefSeq

Specifications

Antigen ITGA2
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 244 G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ITGA2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene ITGA2
Gene Alias BR/CD49B/GPIa/VLA-2/VLAA2
Gene Symbols ITGA2
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular region of human ITGA2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3673
Test Specificity Recognizes the extracellular domain of human alpha2 integrin.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.