Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2, Mouse, Clone: Gi9, Abnova™

Catalog No. 89111963 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-963 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-963 Supplier Abnova Corporation Supplier No. MAB4665

Mouse monoclonal antibody raised against native ITGA2.

This gene product belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane glycoproteins composed of a distinct alpha chain and a common beta chain. They are found on a wide variety of cell types including, T cells, fibroblasts and platelets. Integrins are involved in cell adhesion and also participate in cell-surface mediated signalling. [provided by RefSeq

Specifications

Antigen ITGA2
Applications Flow Cytometry
Classification Monoclonal
Clone Gi9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ITGA2.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (1mg/mL BSA)
Gene ITGA2
Gene Alias BR/CD49B/GPIa/VLA-2/VLAA2
Gene Symbols ITGA2
Host Species Mouse
Immunogen Native purified ITGA2 from isolated human platelets.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3673
Test Specificity The molecular weight of the recognized antigen is 138/115KDa under non-reducing conditions and 145/126KDa under reduced conditions. Recognizes the VLA2 antigen (alpha 2 beta 1 complex) also called Ia/IIa glycoprotein complex or collagen receptor. This antibody reacts with the molecules carrying the Br(a) as well as the Br(b) alloantigen. It reacts with platelets, megakaryocytes and activated T cells. Does not react with B lymphocytes, T lymphocytes, monocytes, granulocytes and red blood cells. This antibody inhibits the adhesion of platelets to collagen.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.