Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2B, Mouse, Clone: P2, Abnova™

Catalog No. 89111713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-713 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-713 Supplier Abnova Corporation Supplier No. MAB4143
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ITGA2B.

ITGA2B encodes integrin alpha chain 2b. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. Alpha chain 2b undergoes post-translational cleavage to yield disulfide-linked light and heavy chains that join with beta 3 to form a fibronectin receptor expressed in platelets that plays a crucial role in coagulation. Mutations that interfere with this role result in thrombasthenia. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling. [provided by RefSeq

Specifications

Antigen ITGA2B
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone P2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ITGA2B.
Dilution Flow Cytometry (2ug/5x106 platelets/test) Immunohistochemistry (Frozen sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (1mg/mL BSA)
Gene ITGA2B
Gene Alias CD41/CD41B/GP2B/GPIIb/GTA/HPA3
Gene Symbols ITGA2B
Host Species Mouse
Immunogen Native purified ITGA2B from human platelets.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Test Specificity Reacts with gpIIb in the intact complex with gpIIIa, but not with IIb or IIIa, separately. Inhibits platelet aggregation induced by thrombin, collagen and ADP. The molecular weight of the recognized complex is 135KDa/95KDa.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.