Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2B/ITGB3, Mouse, Clone: 8, Abnova™

Catalog No. 89112851 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-851 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-851 Supplier Abnova Corporation Supplier No. MAB6393
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against ITGA2B/ITGB3.

Specifications

Antigen ITGA2B/ITGB3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against ITGA2B/ITGB3.
Dilution Western Blot (10-20 ug/mL) ELISA (100ul of 108 washed platelets) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene Alias CD41/CD41B/GP2B/GPIIb/GTA/HPA3
Gene Symbols ITGA2B
Host Species Mouse
Immunogen Native from human platelets suspension.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674/3690
Test Specificity Under reducing conditions this antibody will bind against the human ITGA2B/ITGB3 complex.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.