Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA5, Mouse, Clone: CLB-705, Abnova™

Catalog No. 89112198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-198 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-198 Supplier Abnova Corporation Supplier No. MAB5267

Mouse monoclonal antibody raised against native ITGA5.

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling. [provided by RefSeq

Specifications

Antigen ITGA5
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone CLB-705
Conjugate Unconjugated
Description integrin, alpha 5 (fibronectin receptor, alpha polypeptide)
Formulation In 20mM Tris, 150mM NaCl, pH 8.0 (10mg BSA, 0.001% thimerosal)
Gene ITGA5
Gene Alias CD49e, FNRA, VLA5A
Gene Symbols ITGA5
Host Species Mouse
Immunogen Native purified from U937 cell line.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.