Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA6, Mouse, Clone: 541A11, Abnova™

Catalog No. 89111429 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-429 Supplier Abnova Corporation Supplier No. MAB3462
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ITGA6.

The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen ITGA6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 541A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ITGA6.
Dilution The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene ITGA6
Gene Alias CD49f/DKFZp686J01244/FLJ18737/ITGA6B/VLA-6
Gene Symbols ITGA6
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular region of human ITGA6.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3655
Test Specificity Recognizes the extracellular domain of human alpha6 integrin.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.