Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAM, Mouse, Biotin, Clone: ICRF44, Abnova™

Catalog No. 89112642 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-642 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-642 Supplier Abnova Corporation Supplier No. MAB6031
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ITGAM.

This gene encodes the integrin alpha M chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as macrophage receptor 1 ('Mac-1'), or inactivated-C3b (iC3b) receptor 3 ('CR3'). The alpha M beta 2 integrin is important in the adherence of neutrophils and monocytes to stimulated endothelium, and also in the phagocytosis of complement coated particles. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen ITGAM
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ICRF44
Conjugate Biotin
Description Mouse monoclonal antibody raised against native ITGAM.
Dilution Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene ITGAM
Gene Alias CD11B/CR3A/MAC-1/MAC1A/MGC117044/MO1A/SLEB6
Gene Symbols ITGAM
Host Species Mouse
Immunogen Native purified ITGAM from rheumatoid synovial cells and human monocytes.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity Specificity human CD11b/Mac-1alpha
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.