Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB1, Mouse, Biotin, Clone: MEM-174, Abnova™

Catalog No. 89111887 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-887 Supplier Abnova Corporation Supplier No. MAB4546
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ITGAM.

This gene encodes the integrin alpha M chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as macrophage receptor 1 ('Mac-1'), or inactivated-C3b (iC3b) receptor 3 ('CR3'). The alpha M beta 2 integrin is important in the adherence of neutrophils and monocytes to stimulated endothelium, and also in the phagocytosis of complement coated particles. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen ITGB1
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-174
Conjugate Biotin
Description Mouse monoclonal antibody raised against native ITGAM.
Dilution Flow Cytometry (1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene ITGAM
Gene Alias CD11B/CR3A/MAC-1/MAC1A/MGC117044/MO1A/SLEB6
Gene Symbols ITGAM
Host Species Mouse
Immunogen Native purified ITGAM from human granulocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity This antibody recognizes CD11b antigen (Mac-1 alpha), a 165-170 KDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.