Learn More
ITGAM, Mouse, FITC, Clone: MEM-174, Abnova™
Mouse monoclonal antibody raised against native ITGAM.
Supplier: Abnova Corporation MAB4549
Description
This gene encodes the integrin alpha M chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as macrophage receptor 1 ('Mac-1'), or inactivated-C3b (iC3b) receptor 3 ('CR3'). The alpha M beta 2 integrin is important in the adherence of neutrophils and monocytes to stimulated endothelium, and also in the phagocytosis of complement coated particles. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
ITGAM | |
Monoclonal | |
FITC | |
Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
ITGAM | |
ITGAM | |
Native purified ITGAM from human granulocytes. | |
RUO | |
3684 | |
Human | |
Liquid |
Flow Cytometry | |
MEM-174 | |
Mouse monoclonal antibody raised against native ITGAM. | |
In PBS (0.2% BSA, 0.09% sodium azide) | |
CD11B/CR3A/MAC-1/MAC1A/MGC117044/MO1A/SLEB6 | |
Mouse | |
100 Reactions | |
Primary | |
This antibody recognizes CD11b antigen (Mac-1 alpha), a 165-170 KDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.