Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAV, Mouse, Clone: NKI-M9, Abnova™

Catalog No. 89111368 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-368 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-368 Supplier Abnova Corporation Supplier No. MAB3354

Mouse monoclonal antibody raised against native ITGAV.

ITAGV encodes integrin alpha chain V. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. The I-domain containing integrin alpha V undergoes post-translational cleavage to yield disulfide-linked heavy and light chains, that combine with multiple integrin beta chains to form different integrins. Among the known associating beta chains (beta chains 1,3,5,6, and 8; 'ITGB1', 'ITGB3', 'ITGB5', 'ITGB6', and 'ITGB8'), each can interact with extracellular matrix ligands; the alpha V beta 3 integrin, perhaps the most studied of these, is referred to as the Vitronectin receptor (VNR). In addition to adhesion, many integrins are known to facilitate signal transduction. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Specifications

Antigen ITGAV
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone NKI-M9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ITGAV.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris, 150mM NaCl, pH 8.0 (0.001% thimerosal)
Gene ITGAV
Gene Alias CD51/DKFZp686A08142/MSK8/VNRA
Gene Symbols ITGAV
Host Species Mouse
Immunogen Native purified ITGAV from human melanoma cells.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Test Specificity This antibody is directed against the CD51 antigen, alpha chain of vitronectin receptor.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.