Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAX, Mouse, Clone: FK24, Abnova™

Catalog No. 89135205 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-205 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-205 Supplier Abnova Corporation Supplier No. MAB2530
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ITGAX.

Specifications

Antigen ITGAX
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone FK24
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ITGAX.
Dilution Immunofluorescence (1:5) Immunocytochemistry (1:5) The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.01% sodium azide)
Gene Alias CD11C/SLEB6
Gene Symbols ITGAX
Host Species Mouse
Immunogen AML-blasts.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3687
Test Specificity This antibody reacts with the alpha chain the 150 kD molecule of the CD11c/CD18 Integrin complex (g150/95) which is expressed on monocytes and in a lower density on granulocytes and NK-cells of peripheral blood. It also reacts with many myelomonocytic and monocytic leukemias and (weakly) with part of the T8 positive cells (OKM1-like).
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.