Learn More
ITGAX, Mouse, PerCP, Clone: BU15, Abnova™
Mouse monoclonal antibody raised against native ITGAX.
Supplier: Abnova Corporation MAB4353
Description
This gene encodes the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. [provided by RefSeq
Specifications
| ITGAX | |
| Monoclonal | |
| PerCP | |
| Flow Cytometry (10 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
| ITGAX | |
| ITGAX | |
| Native purified ITGAX from human dendritic cells of synovial fluid. | |
| RUO | |
| 3687 | |
| Canine, Human, Monkey | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| BU15 | |
| Mouse monoclonal antibody raised against native ITGAX. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| CD11C/SLEB6 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| This antibody reacts with ITGAX, a 150 KDa integrin expressed mainly on dendritic cells and tissue macrophages. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.