Learn More
ITGB1, Mouse, Clone: k2D5, Abnova™
Mouse monoclonal antibody raised against partial recombinant ITGB1.
Supplier: Abnova Corporation MAB1129
Description
Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene. [provided by RefSeq
Specifications
ITGB1 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
ITGB1 | |
CD29/FNRB/GPIIA/MDF2/MSK12/VLA-BETA/VLAB | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
k2D5 | |
Mouse monoclonal antibody raised against partial recombinant ITGB1. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
NM_002211 | |
ITGB1 | |
Recombinant protein corresponding to amino acids 34-141 of human ITGB1. | |
RUO | |
3688 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.