Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB1, Mouse, FITC, Clone: MEM-101A, Abnova™

Catalog No. 89111891 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-891 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-891 Supplier Abnova Corporation Supplier No. MAB4550
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ITGB1.

Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen ITGB1
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-101A
Conjugate FITC
Description Mouse monoclonal antibody raised against native ITGB1.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene ITGB1
Gene Alias CD29/FNRB/GPIIA/MDF2/MSK12/VLA-BETA/VLAB
Gene Symbols ITGB1
Host Species Mouse
Immunogen Native purified ITGB1 from Raji Burkitt's lymphoma cell line.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3688
Test Specificity This antibody reacts with CD29 (integrin beta1 chain), a 130 KDa single chain type I glycoprotein expressed as a heterodimer (non-covalently associated with the integrin alpha subunits 1-6).
Target Species Canine, Human, Pig
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.