Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itgb3, Mouse, Clone: MHF4, Abnova™

Catalog No. 89113126 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-126 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-126 Supplier Abnova Corporation Supplier No. MAB7098

Mouse monoclonal antibody raised against native Itgb3.

Specifications

Antigen Itgb3
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone MHF4
Conjugate Unconjugated
Description integrin beta 3
Formulation In PBS
Gene Itgb3
Gene Symbols Itgb3
Host Species Mouse
Immunogen Native purified Itgb3 from a bone cell suspension containing osteoclasts, osteoblasts and bone marrow cells obtained from the long bones of newborn Sprague Dawley rats.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29302
Test Specificity MHF4 may be used to identify osteoclasts in tissue sections and bone cell suspensions. Not yet tested in other species. Not tested in other applications.
Target Species Human, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.