Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITK, Mouse, Clone: 5G6, Abnova™

Catalog No. 89108906 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-906 Supplier Abnova Corporation Supplier No. MAB10704
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ITK.

This gene encodes an intracellular tyrosine kinase expressed in T-cells. The protein contains both SH2 and SH3 domains which are often found in intracellular kinases. It is thought to play a role in T-cell proliferation and differentiation. [provided by RefSeq

Specifications

Antigen ITK
Applications ELISA, Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ITK.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene ITK
Gene Alias EMT/LYK/MGC126257/MGC126258/PSCTK2
Gene Symbols ITK
Host Species Mouse
Immunogen Recombinant protein corresponding to human ITK.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3702
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.