Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IVL, Mouse, Clone: SY5, Abnova™

Catalog No. 89109528 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-528 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-528 Supplier Abnova Corporation Supplier No. MAB11366
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against IVL.

Involucrin, a component of the keratinocyte crosslinked envelope, is found in the cytoplasm and crosslinked to membrane proteins by transglutaminase. This gene is mapped to 1q21, among calpactin I light chain, trichohyalin, profillaggrin, loricrin, and calcyclin. [provided by RefSeq

Specifications

Antigen IVL
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SY5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against IVL.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (1-2 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determine
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene IVL
Gene Symbols IVL
Host Species Mouse
Immunogen Purified IVL from human keratinocytes.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Canine, Gorilla, Human, Monkey, Owl, Pig
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.