Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK2, Mouse, Clone: 691R5, Abnova™

Catalog No. 89110704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-704 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-704 Supplier Abnova Corporation Supplier No. MAB1906

Mouse monoclonal antibody raised against full-length recombinant JAK2.

This gene product is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways. It has been found to be constituitively associated with the prolactin receptor and is required for responses to gamma interferon. Mice that do not express an active protein for this gene exhibit embryonic lethality associated with the absence of definitive erythropoiesis. [provided by RefSeq

Specifications

Antigen JAK2
Applications Western Blot
Classification Monoclonal
Clone 691R5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant JAK2.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (1.0% BSA, 0.08% sodium azide)
Gene JAK2
Gene Alias JTK10
Gene Symbols JAK2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human JAK2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3717
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.