Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK3, Mouse, Clone: 5H2, Abnova™

Catalog No. 89108907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-907 Supplier Abnova Corporation Supplier No. MAB10705
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant JAK3.

The protein encoded by this gene is a member of the Janus kinase (JAK) family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction. It is predominantly expressed in immune cells and transduces a signal in response to its activation via tyrosine phosphorylation by interleukin receptors. Mutations in this gene are associated with autosomal SCID (severe combined immunodeficiency disease). [provided by RefSeq

Specifications

Antigen JAK3
Applications ELISA, Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant JAK3.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene JAK3
Gene Alias JAK-3/JAK3_HUMAN/JAKL/L-JAK/LJAK
Gene Symbols JAK3
Host Species Mouse
Immunogen Recombinant protein corresponding to human JAK3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3718
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.