Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-JARID1D/KDM5D, Clone: 7F10.4,

Catalog No. MABE326MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE326MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE326MI Supplier MilliporeSigma™ Supplier No. MABE326
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects JARID1D/KDM5D clone: 7F10.4 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

Lysine-specific demethylase 5D (UniProt Q9BY66; also known as H-Y, Histone demethylase JARID1D, Histocompatibility Y antigen, Human homolog of selected mouse cDNA on Y, Jumonji/ARID domain-containing protein 1D, Jumonji AT rich interactive domain 1D (RBP2-like), Protein SmcY, SMC homolog Y chromosome, Smcy homolog Y-linked) is encoded by the KDM5D (also known as HY, HYA, JARID1D, KIAA0234, SMCY) gene (Gene ID 8284) in human. JARID1D/KDM5D is a histone demethylase that specifically catalyzes the demethylation of tri- and di-methylated Lys4 of histone H3 (H3K4m3 and H3K4m2), but not methylated H3K9, H3K27, H3K36, H3K79, H4K20, or monomethylated H3K4. The KDM5D gene is mapped to the azoospermia factor (AZF) region known to be involved in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JARID1D/KDM5D
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7F10.4
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9BY66
Gene Symbols KDM5D; HY; HYA; JARID1D; KIAA0234; SMCY
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human JARID1D/KDM5D.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001140177
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.