Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD1A, Mouse, Clone: 2E1, Abnova™

Catalog No. 89108909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-909 Supplier Abnova Corporation Supplier No. MAB10707
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant JMJD1A.

This gene encodes a zinc finger protein that contains a jumonji domain and may play a role in hormone-dependent transcriptional activation. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen JMJD1A
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2E1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant JMJD1A.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene JMJD1A
Gene Alias DKFZp686A24246/DKFZp686P07111/JHDM2A/JHMD2A/JMJD1/KDM3A/KIAA0742/TSGA
Gene Symbols JMJD1A
Host Species Mouse
Immunogen Recombinant protein corresponding to human JMJD1A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55818
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.