Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JUN, Mouse, Clone: 4H9, Abnova™

Catalog No. 89108702 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-702 Supplier Abnova Corporation Supplier No. MAB10515
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant JUN.

This gene is the putative transforming gene of avian sarcoma virus 17. It encodes a protein which is highly similar to the viral protein, and which interacts directly with specific target DNA sequences to regulate gene expression. This gene is intronless and is mapped to 1p32-p31, a chromosomal region involved in both translocations and deletions in human malignancies. [provided by RefSeq

Specifications

Antigen JUN
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4H9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant JUN.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene JUN
Gene Alias AP-1/AP1/c-Jun
Gene Symbols JUN
Host Species Mouse
Immunogen Recombinant protein corresponding to human JUN.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3725
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.