Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa light chains, Mouse, Clone: TB28-2, Abnova™

Catalog No. 89108415 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-415 Supplier Abnova Corporation Supplier No. MAB10167
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Kappa light chains.

Specifications

Antigen Kappa light chains
Applications Flow Cytometry
Classification Monoclonal
Clone TB28-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Kappa light chains.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene IGK@
Gene Alias FLJ26296/IGK/IGKC/MGC22645/MGC27376/MGC40426/MGC71990
Gene Symbols IGK@
Host Species Mouse
Immunogen Human IgG-kappa myeloma protein.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50802
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.