Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT5, Mouse, Clone: 3F9, Abnova™

Catalog No. 89108584 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-584 Supplier Abnova Corporation Supplier No. MAB10398
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant KAT5.

The protein encoded by this gene belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein. HATs play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins. This protein is a histone acetylase that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen KAT5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant KAT5.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene KAT5
Gene Alias ESA1/HTATIP/HTATIP1/PLIP/TIP/TIP60/cPLA2
Gene Symbols KAT5
Host Species Mouse
Immunogen Recombinant protein corresponding to human KAT5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10524
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.