Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcna1, Mouse, Clone: S20-78, Abnova™

Catalog No. 89113505 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-505 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-505 Supplier Abnova Corporation Supplier No. MAB8574
This item is not returnable. View return policy

Mouse monoclonal antibody raised against synthetic peptide of Kcna1.

Specifications

Antigen Kcna1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S20-78
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Kcna1.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Kcna1
Gene Alias Kcna/Kcpvd/Kv1.1
Gene Symbols Kcna1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 458-476 of rat Kcna1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24520
Test Specificity This antibody is specific to Kcna1.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.