Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcna3, Mouse, R-PE, Clone: S23-27, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of Kcna3.
Supplier: Abnova Corporation MAB11672
Description
Sequence: GGMNHSAFPQTPFKTGNSYATCSpecifications
Kcna3 | |
Monoclonal | |
R-PE | |
Immunohistochemistry (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Kcna3 | |
Mouse | |
Protein G purification | |
RUO | |
29731 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Immunofluorescence, Immunohistochemistry, Western Blot | |
S23-27 | |
Mouse monoclonal antibody raised against synthetic peptide of Kcna3. | |
In PBS pH 7.4 (50% glycerol and 0.09% sodium azide) | |
Kcna3 | |
A synthetic peptide corresponding to amino acids 485-506 of rat Kcna3. | |
100 μg | |
Primary | |
Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction