Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcna6, Mouse, Clone: S19-36, Abnova™

Catalog No. 89109952 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-952 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-952 Supplier Abnova Corporation Supplier No. MAB11818

Mouse monoclonal antibody raised against synthetic peptide of Kcna6.

Sequence: RERRSSYLPTPHRAYAEKR

Specifications

Antigen Kcna6
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone S19-36
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Kcna6.
Dilution Western Blot (1 ug/mL) Immunocytochemistry (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) Immunohistochemistry (0.1-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Kcna6
Gene Alias Kv1.6
Gene Symbols Kcna6
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids of 507-525 at cytoplasmic C-terminus of rat Kcna6.
Purification Method PEG Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64358
Test Specificity Does not cross react with other Kv1 channels.
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.