Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcnab1, Mouse, Clone: S40-17, Abnova™
Mouse monoclonal antibody raised against full-length recombinant Kcnab1.
Supplier: Abnova Corporation MAB8579
Specifications
Kcnab1 | |
Monoclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user. | |
Kcnab1 | |
Kcnab1 | |
Recombinant protein corresponding to full length of rat Kcnab1. | |
RUO | |
29737 | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Western Blot | |
S40-17 | |
Mouse monoclonal antibody raised against full length recombinant Kcnab1. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
KvBeta3/MGC105463 | |
Mouse | |
100 μg | |
Primary | |
This antibody is specific to Kcnab1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction