Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnab2, Mouse, Clone: S17-70, Abnova™

Catalog No. 89113504 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-504 Supplier Abnova Corporation Supplier No. MAB8573

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against full-length recombinant Kcnab2.

Specifications

Antigen Kcnab2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S17-70
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Kcnab2.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Kcnab2
Gene Symbols Kcnab2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of rat Kcnab2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29738
Test Specificity This antibody is specific to Kcnab2.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.