Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcnb2, Mouse, FITC, Clone: S37-89, Abnova™
Mouse monoclonal antibody raised against partial recombinant Kcnb2.
Supplier: Abnova Corporation MAB11848
Specifications
Kcnb2 | |
Monoclonal | |
FITC | |
Western Blot (1 ug/mL) Immunocytochemistry (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) Immunohistochemistry (0.1-1 ug/mL) The optimal working dilution should be determined by the end user. | |
Kcnb2 | |
Kcnb2 | |
Recombinant fusion protein corresponding to amino acids of 1-61 of rat Kcnb2. | |
100 μg | |
Primary | |
Rat | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
S37-89 | |
Mouse monoclonal antibody raised against partial recombinant Kcnb2. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
Kv2.2 | |
Mouse | |
Protein G Purification | |
RUO | |
117105 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction