Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnj2, Mouse, Clone: S112B-14, Abnova™

Catalog No. 89112983 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-983 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-983 Supplier Abnova Corporation Supplier No. MAB6657
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Kcnj2.

Specifications

Antigen Kcnj2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S112B-14
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Kcnj2.
Dilution Western Blot (1-10 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Kcnj2
Gene Alias IRK1/Kcnf1/Kir2.1
Gene Symbols Kcnj2
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 390-411 of mouse Kcnj2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16518
Test Specificity Detects ∼55KDa. No cross-reactivity against Kir2.2 or Kir2.3.
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.