Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNQ1, Mouse, Clone: S37A-10, Abnova™

Catalog No. 89112980 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-980 Supplier Abnova Corporation Supplier No. MAB6654
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant KCNQ1.

This gene encodes a protein for a voltage-gated potassium channel required for the repolarization phase of the cardiac action potential. The gene product can form heteromultimers with two other potassium channel proteins, KCNE1 and KCNE3. Mutations in this gene are associated with hereditary long QT syndrome, Romano-Ward syndrome, Jervell and Lange-Nielsen syndrome and familial atrial fibrillation. The gene is located in a region of chromosome 11 that contains a large number of contiguous genes that are abnormally imprinted in cancer and the Beckwith-Wiedemann syndrome. Two alternative transcripts encoding distinct isoforms have been described. [provided by RefSeq

Specifications

Antigen KCNQ1
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S37A-10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant KCNQ1.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1.0-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene KCNQ1
Gene Alias ATFB1/FLJ26167/JLNS1/KCNA8/KCNA9/KVLQT1/Kv1.9/Kv7.1/LQT/LQT1/RWS/SQT2/WRS
Gene Symbols KCNQ1
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to amino acids 2-101 of human KCNQ1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3784
Test Specificity Detects ∼75KDa.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.