Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD15, Mouse, Clone: AT4C3, Abnova™

Catalog No. 89113469 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-469 Supplier Abnova Corporation Supplier No. MAB8482
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant KCTD15.

Specifications

Antigen KCTD15
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AT4C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant KCTD15.
Dilution Western Blot (1:250-1:500) Immunofluorescence (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene KCTD15
Gene Alias MGC25497/MGC2628
Gene Symbols KCTD15
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human KCTD15.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79047
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3 γ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.