Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDEL, Mouse, Clone: 10C3, Abnova™

Catalog No. 89110880 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-880 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-880 Supplier Abnova Corporation Supplier No. MAB2193
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of KDEL.

Specifications

Antigen KDEL
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 10C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of KDEL.
Dilution Western Blot (4 ug/mL) Immunocytochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.1mM PMSF)
Gene Hspa5
Gene Alias BIP
Gene Symbols Hspa5
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 649-654 of rat Hspa5.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25617
Test Specificity This antibody recognizes proteins containing the KDEL sequence in mammals, birds, insects and plants. Cross-reacts with yeast proteins containing the HDEL sequence.
Target Species Avian, Insect, Rat, Yeast
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.