Learn More
KDM1A Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant KDM1A.
Supplier: Abnova Corporation MAB8721
Description
This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
KDM1A | |
Monoclonal | |
Unconjugated | |
Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user. | |
AOF2 | |
BHC110/KDM1/KIAA0601/LSD1 | |
Mouse | |
100 μg | |
Primary | |
This antibody is specific to KDM1A. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
1 mg/mL | |
Mouse monoclonal antibody raised against partial recombinant KDM1A. | |
In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol) | |
NM_001009999 | |
AOF2 | |
Recombinant protein corresponding to N-terminus residues of human KDM1A. | |
RUO | |
23028 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.